Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lin7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011698.1 | NP_035828.1 | 2, 3 | 159 | ||
GenBank mRNA | AF087694.1 | - | 2, 3 | 166 | ||
AF173082.1 | - | 2, 3 | 159 | |||
AK019299.1 | - | 2, 3 | 164 | |||
BC031780.1 | - | 2, 3 | 178 | 4164883 |