Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scarf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153790.2 | NP_722485.1 | 4 | 618 | ||
GenBank mRNA | AB024432.1 | - | 4 | 585 | ||
AF522197.1 | - | 4 | 619 | |||
AK142378.1 | - | 4 | 586 | |||
AK143208.1 | - | 4 | 461 | |||
AK214903.1 | - | 1 | 61 | |||
BC059226.1 | - | 4 | 550 | |||
BC060084.1 | - | 4 | 550 | |||
BC139758.1 | - | 4 | 571 | 40047931 |