Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnt7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009527.3 | NP_033553.2 | 2 | 524 | ||
GenBank mRNA | AK004683.1 | - | 2 | 525 | ||
BC049093.1 | - | 2 | 518 | 6415801 | ||
BC057586.1 | - | 2 | 518 | 6415801 | ||
M89801.1 | - | 2 | 264 |