Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp160 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145483.2 | NP_663458.2 | 5 | 1521 | ||
GenBank mRNA | AK143313.1 | - | 5 | 1522 | ||
AK173216.1 | - | 5 | 1553 | |||
BC015291.1 | - | 5 | 1487 | 4242025 |