Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Zfp81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207541.1 | NP_997424.1 | 4 | 740 | ||
GenBank mRNA | AK079439.1 | - | 1 | 67 | ||
AK135853.1 | - | 4 | 741 | |||
BC026640.1 | - | 1 | 163 | |||
BC117831.1 | - | 4 | 690 | 40092935 | ||
BC117832.1 | - | 4 | 692 | 40092936 |