Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146097.3 | NP_666209.1 | 7, 8 | 644 | ||
GenBank mRNA | AK076027.1 | - | 7, 8 | 599 | ||
AK132291.1 | - | 7, 8 | 630 | |||
BC018472.1 | - | 7, 8 | 586 | 3661842 | ||
BC053745.1 | - | 7, 8 | 679 | 6505520 |