Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lzts2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130525.1 | NP_001123997.1 | 3 | 681 | ||
NM_001130526.1 | NP_001123998.1 | 3 | 673 | |||
NM_145503.2 | NP_663478.2 | 3 | 725 | |||
GenBank mRNA | AK081367.1 | - | 3 | 598 | ||
AK129446.1 | - | 2 | 590 | |||
AK144154.1 | - | 3 | 692 | |||
AK169936.1 | - | 3 | 714 | |||
BC014695.1 | - | 2 | 574 | 3157493 | ||
BC092263.1 | - | 3 | 705 | 2654420 |