Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ablim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103177.2 | NP_001096647.1 | 10, 11 | 1362 | ||
NM_001103178.2 | NP_001096648.1 | 7, 8 | 784 | |||
NM_001290813.1 | NP_001277742.1 | 10, 11 | 1348 | |||
NM_001290815.1 | NP_001277744.1 | 10, 11 | 1362 | |||
NM_001290816.1 | NP_001277745.1 | 10, 11 | 1362 | |||
NM_178688.3 | NP_848803.3 | 12, 13 | 1480 | |||
GenBank mRNA | AF404774.1 | - | 12, 13 | 1480 | ||
AF404775.1 | - | 10, 11 | 1122 | |||
AF404776.1 | - | 7, 8 | 744 | |||
AK079331.1 | - | 6, 7 | 602 | |||
AK122196.1 | - | 10, 11 | 1362 | |||
AK143907.1 | - | 10, 11 | 1132 | |||
AK171860.1 | - | 1259 | ||||
BC032216.1 | - | 1, 2 | 157 | |||
BC141083.1 | - | 10, 11 | 1342 | 9055695 |