Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zpr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011752.2 | NP_035882.1 | 12 | 1248 | ||
GenBank mRNA | AB041612.1 | - | 12 | 1131 | ||
AK016551.1 | - | 6 | 415 | |||
AK044472.1 | - | 12 | 1137 | |||
AK167849.1 | - | 12 | 1119 | |||
AK168061.1 | - | 12 | 1090 | |||
AK170372.1 | - | 3 | 1911 | |||
AK194385.1 | - | 3 | 209 | |||
BC021397.1 | - | 12 | 1131 | 5067274 |