Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146117.2 | NP_666229.1 | 1 | 360 | ||
GenBank mRNA | AK137758.1 | - | 1 | 356 | ||
AK142669.1 | - | 1 | 348 | |||
BC004853.1 | - | 1 | 355 | 3154380 | ||
BC024531.1 | - | 1 | 330 | 4987362 | ||
BC024539.1 | - | 1 | 324 | 4988839 |