Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110512.1 | NP_001103982.1 | 7 | 699 | ||
NM_145524.3 | NP_663499.2 | 8 | 1163 |