Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cerkl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048176.1 | NP_001041641.1 | 12 | 1503 | ||
GenBank mRNA | BC046474.1 | - | 12 | 1609 | ||
CJ235603.1 | - | 6 | 434 | |||
HQ426680.1 | - | 12 | 1503 | |||
HQ426681.1 | - | 10 | 1296 | |||
HQ426682.1 | - | 9 | 1164 | |||
HQ426683.1 | - | 8 | 921 | |||
HQ426684.1 | - | 11 | 1371 | |||
HQ426685.1 | - | 11 | 1443 | |||
HQ426686.1 | - | 11 | 1360 | |||
HQ426687.1 | - | 12 | 1634 | |||
HQ426688.1 | - | 12 | 1650 | |||
HQ426689.1 | - | 11 | 1518 | |||
HQ426690.1 | - | 12 | 1720 | |||
HQ426691.1 | - | 1302 | ||||
HQ426692.1 | - | 1433 | ||||
HQ426693.1 | - | 1343 | ||||
HQ426694.1 | - | 1256 | ||||
HQ426695.1 | - | 1475 | ||||
HQ426696.1 | - | 1415 | ||||
HQ426697.1 | - | 1560 | ||||
HQ426698.1 | - | 1268 | ||||
HQ426699.1 | - | 1332 | ||||
HQ426700.1 | - | 1606 | ||||
HQ426701.1 | - | 1622 | ||||
HQ426702.1 | - | 11 | 1085 | |||
HQ426703.1 | - | 9 | 878 | |||
HQ426704.1 | - | 12 | 1242 | |||
HQ426705.1 | - | 10 | 942 | |||
HQ426706.1 | - | 10 | 1029 | |||
HQ426707.1 | - | 10 | 1176 | |||
HQ426708.1 | - | 6 | 539 | |||
HQ426709.1 | - | 9 | 934 | |||
HQ426710.1 | - | 8 | 874 | |||
HQ426711.1 | - | 9 | 1081 |