Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cerkl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048176.1 | NP_001041641.1 | 2 | 492 | ||
GenBank mRNA | BC046474.1 | - | 2 | 598 | ||
BY742285.1 | - | 2 | 502 | |||
HQ426680.1 | - | 2 | 492 | |||
HQ426681.1 | - | 2 | 492 | |||
HQ426682.1 | - | 2 | 492 | |||
HQ426684.1 | - | 2 | 492 | |||
HQ426685.1 | - | 2 | 492 | |||
HQ426686.1 | - | 2 | 492 | |||
HQ426687.1 | - | 2 | 623 | |||
HQ426688.1 | - | 2 | 492 | |||
HQ426689.1 | - | 2 | 492 | |||
HQ426690.1 | - | 2 | 492 | |||
HQ426691.1 | - | 291 | ||||
HQ426692.1 | - | 422 | ||||
HQ426693.1 | - | 332 | ||||
HQ426694.1 | - | 291 | ||||
HQ426695.1 | - | 464 | ||||
HQ426696.1 | - | 464 | ||||
HQ426697.1 | - | 464 | ||||
HQ426698.1 | - | 464 | ||||
HQ426699.1 | - | 464 | ||||
HQ426700.1 | - | 595 | ||||
HQ426701.1 | - | 464 |