Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp217 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033299.3 | NP_001028471.1 | 2 | 737 | ||
NM_001159683.1 | NP_001153155.1 | 3 | 722 | |||
GenBank mRNA | AK139547.1 | - | 3 | 724 | ||
AK156475.1 | - | 2 | 738 | |||
AK163381.1 | - | 3 | 727 | |||
BC116329.1 | - | 1 | 216 | 40096805 | ||
BC116330.1 | - | 1 | 216 | 40096806 |