Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tram1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146140.3 | NP_666252.1 | 1 | 1066 | ||
GenBank mRNA | AK044115.1 | - | 1 | 993 | ||
AK048118.1 | - | 1016 | ||||
AK052946.1 | - | 1 | 1038 | |||
AK083053.1 | - | 1 | 995 | |||
BC024829.1 | - | 1 | 993 | 5359105 | ||
BC027120.1 | - | 1 | 993 | 5359106 |