Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifna16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206867.1 | NP_996750.1 | 1 | 358 | ||
GenBank mRNA | BC114392.1 | - | 1 | 357 |