Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zcchc11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175472.3 | NP_780681.2 | 3 | 885 | ||
GenBank mRNA | AF194972.1 | - | 2 | 650 | ||
AK031299.1 | - | 1 | 278 | |||
AK041821.1 | - | 3 | 891 | |||
AK220327.1 | - | 1 | 133 | |||
BC038132.1 | - | 2 | 646 | |||
BC145507.1 | - | 2 | 536 | |||
BC150791.1 | - | 2 | 536 | 9087702 |