Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfyve9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183300.2 | NP_899123.2 | 15 | 1891 | ||
GenBank mRNA | BC038255.1 | - | 4 | 386 | ||
BC046341.1 | - | 5 | 527 | |||
BC058416.1 | - | 5 | 451 | |||
BC079578.1 | - | 5 | 459 | |||
DQ303413.1 | - | 15 | 1891 |