Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Szt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198170.4 | NP_937813.3 | 6 | 779 | ||
GenBank mRNA | FJ998170.1 | - | 6 | 779 | ||
GU433214.1 | - | 6 | 705 |