Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Yrdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153566.2 | NP_705794.2 | 4 | 737 | ||
GenBank mRNA | AK153627.1 | - | 4 | 737 | ||
AK219737.1 | - | 2 | 202 | |||
AY283537.1 | - | 4 | 713 | |||
BC023823.1 | - | 4 | 723 | 5343817 |