Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Megf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162977.1 | NP_001156449.1 | 23 | 3190 | ||
GenBank mRNA | AK134303.1 | - | 10 | 1184 | ||
AK173032.1 | - | 6 | 669 | |||
BC031402.1 | - | 3 | 228 | |||
BC039980.1 | - | 3 | 239 | |||
BC058571.1 | - | 2 | 132 |