Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4931409K22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177676.6 | NP_808344.3 | 17 | 2197 | ||
GenBank mRNA | AK029839.1 | - | 16 | 2629 | ||
AV271477.2 | - | 1 | 143 | |||
BC137999.1 | - | 13 | 1918 | 8861018 | ||
BC145792.1 | - | 13 | 1920 | 40131095 |