Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033310.3 | NP_001028482.2 | 1 | 357 | ||
NM_001163456.1 | NP_001156928.1 | 1 | 357 | |||
GenBank mRNA | AK156053.1 | - | 1 | 326 | ||
BC038311.1 | - | 1 | 127 | |||
BC138972.1 | - | 1 | 335 | 8861994 | ||
BC145479.1 | - | 1 | 335 | 9054042 | ||
BC145486.1 | - | 1 | 335 | |||
BC145728.1 | - | 1 | 335 | 40131031 | ||
CJ085333.1 | - | 1 | 301 | |||
CX225681.1 | - | 1 | 284 | |||
DN174415.1 | - | 1 | 332 |