Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Suv420h2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115018.1 | NP_001108490.1 | 2 | 180 | ||
NM_146177.2 | NP_666289.2 | 2 | 238 | |||
GenBank mRNA | AK137449.1 | - | 2 | 179 | ||
AK154848.1 | - | 2 | 778 | |||
AY555193.1 | - | 2 | 192 | |||
BC024816.1 | - | 2 | 177 | 5359433 | ||
BY196125.1 | - | 2 | 180 | |||
CF731217.1 | - | 2 | 221 |