Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem41b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153525.5 | NP_705745.3 | 2 | 304 | ||
GenBank mRNA | AK029327.1 | - | 2 | 305 | ||
AK033997.1 | - | 2 | 334 | |||
AK035266.1 | - | 3 | 149 | |||
AK038953.1 | - | 2 | 264 | |||
AK053588.1 | - | 2 | 305 | |||
AK075726.1 | - | 2 | 304 | |||
AK078357.1 | - | 2 | 305 | |||
AK090100.1 | - | 2 | 306 | |||
AK139961.1 | - | 2 | 305 | |||
AK150353.1 | - | 2 | 360 | |||
AK153048.1 | - | 2 | 360 | |||
AK162557.1 | - | 2 | 334 | |||
AK165226.1 | - | 2 | 305 | |||
AK166427.1 | - | 2 | 305 | |||
AK166455.1 | - | 2 | 305 | |||
AK167021.1 | - | 2 | 205 | |||
AK172876.1 | - | 2 | 323 | |||
BC026515.1 | - | 2 | 328 | 4910818 | ||
BC027103.1 | - | 2 | 305 | 5360942 | ||
BC086759.1 | - | 757 |