Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031814.1 | NP_001026984.1 | 28, 29 | 4462 | ||
GenBank mRNA | AK052331.1 | - | 4, 5 | 506 |