Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Armc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146205.2 | NP_666317.2 | 3 | 1929 | ||
GenBank mRNA | AK030000.1 | - | 3 | 786 | ||
AK131111.1 | - | 3 | 1929 | |||
BC032200.1 | - | 3 | 1274 | 5342166 | ||
BC089363.1 | - | 3 | 1170 | 5720275 |