Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027626.1 | NP_081902.1 | 2, 3 | 382 | ||
NM_030263.5 | NP_084539.2 | 7, 8 | 724 | |||
NM_177698.4 | NP_808366.2 | 10, 11 | 2488 | |||
GenBank mRNA | AB220685.1 | - | 10, 11 | 2488 | ||
AK031477.1 | - | 2, 3 | 325 | |||
AK122405.1 | - | 1, 2 | 38 | |||
AK147606.1 | - | 2, 3 | 383 | |||
AK161498.1 | - | 7, 8 | 710 | |||
BC052365.1 | - | 7, 8 | 688 | |||
BC131913.1 | - | 2, 3 | 294 | |||
BC145641.1 | - | 2, 3 | 294 |