Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnot1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205226.1 | NP_001192155.1 | 5 | 647 | ||
NM_153164.3 | NP_694804.3 | 5 | 653 | |||
NM_178078.2 | NP_835179.1 | 5 | 653 | |||
GenBank mRNA | AK086041.1 | - | 5 | 604 | ||
AK169894.1 | - | 5 | 657 | |||
BC058249.1 | - | 5 | 588 | |||
BC157948.1 | - | 5 | 566 | 9088123 | ||
BC158073.1 | - | 5 | 572 | 9007370 | ||
BC172105.1 | - | 5 | 572 | 9054391 |