Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp612 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175480.4 | NP_780689.3 | 5 | 851 | ||
GenBank mRNA | AK046220.1 | - | 5 | 776 | ||
AK133449.1 | - | 5 | 869 | |||
BC004576.1 | - | 1 | 212 | |||
BC048413.1 | - | 5 | 823 | 6488946 |