Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: 6430548M08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163760.1 | NP_001157232.1 | 6 | 903 | ||
NM_001163761.1 | NP_001157233.1 | 6 | 924 | |||
NM_001163762.1 | NP_001157234.1 | 6 | 924 | |||
NM_172286.4 | NP_758490.3 | 6 | 924 | |||
GenBank mRNA | AK032447.1 | - | 6 | 925 | ||
AK044754.1 | - | 5 | 733 | |||
AK046401.1 | - | 6 | 926 | |||
AK051096.1 | - | 6 | 926 | |||
AK122299.1 | - | 6 | 910 | |||
AK133157.1 | - | 6 | 922 | |||
AK172108.1 | - | 6 | 925 | |||
BC027126.1 | - | 6 | 903 | 5359842 |