Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppan | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145610.2 | NP_663585.2 | 8 | 852 | ||
GenBank mRNA | AK039027.1 | - | 8 | 757 | ||
AK085695.1 | - | 1 | 2847 | |||
AK171672.1 | - | 4 | 518 | |||
AK172615.1 | - | 4 | 518 | |||
BC014688.1 | - | 8 | 770 | 4489442 | ||
BC059277.1 | - | 7 | 936 |