Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ash2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080793.2 | NP_001074262.1 | 10 | 946 | ||
NM_001286207.1 | NP_001273136.1 | 10 | 919 | |||
NM_011791.3 | NP_035921.2 | 10 | 1036 | |||
GenBank mRNA | AB020983.2 | - | 10 | 1018 | ||
AK087934.1 | - | 10 | 1011 | |||
AK132471.1 | - | 9 | 942 | |||
AK145576.1 | - | 10 | 911 | |||
AK145790.1 | - | 10 | 923 | |||
AK145967.1 | - | 10 | 890 | |||
AK146254.1 | - | 10 | 948 | |||
AK146599.1 | - | 10 | 920 | |||
AK146756.1 | - | 10 | 911 | |||
AK146938.1 | - | 10 | 1013 | |||
AK180458.1 | - | 2 | 156 | |||
AK183467.1 | - | 5 | 376 | |||
BC012957.1 | - | 10 | 1023 | 4211433 |