Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nek3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162947.1 | NP_001156419.1 | 13 | 1323 | ||
NM_011848.4 | NP_035978.2 | 14 | 1375 | |||
GenBank mRNA | AF093416.1 | - | 13 | 1328 | ||
AF099066.1 | - | 14 | 1344 | |||
BC005411.1 | - | 14 | 1352 | 3590866 |