Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mief1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178719.5 | NP_848834.2 | 6 | 1133 | ||
GenBank mRNA | AK038466.1 | - | 6 | 1106 | ||
AK041184.1 | - | 6 | 1026 | |||
AK044773.1 | - | 7 | 1566 | |||
AK050546.1 | - | 6 | 1094 | |||
AK080375.1 | - | 6 | 1079 | |||
AK163389.1 | - | 7 | 1234 | |||
BC094597.1 | - | 6 | 1065 | |||
BC137633.1 | - | 5 | 1037 | 8860652 |