Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tenm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011858.3 | NP_035988.2 | 31 | 6388 | ||
GenBank mRNA | AB025413.1 | - | 29 | 6221 | ||
AF059485.1 | - | 31 | 6778 | |||
AK122490.1 | - | 5 | 1315 | |||
AK147329.1 | - | 31 | 6389 | |||
AK147579.1 | - | 31 | 6688 |