Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Adamts4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172845.2 | NP_766433.1 | 9 | 2890 | ||
GenBank mRNA | AK028364.1 | - | 2891 | |||
AK076295.1 | - | 8 | 2265 | |||
AK083534.1 | - | 9 | 2890 | |||
AK156429.1 | - | 7 | 1473 | |||
AK157287.1 | - | 2891 | ||||
AK164900.1 | - | 9 | 2886 | |||
AK205486.1 | - | 1 | 87 | |||
AK216415.1 | - | 1 | 87 |