Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011906.2 | NP_036036.2 | 12 | 1516 | ||
GenBank mRNA | AF051098.1 | - | 12 | 1324 | ||
AK155116.1 | - | 12 | 1459 | |||
AK164913.1 | - | 12 | 1273 | |||
AK172045.1 | - | 12 | 1447 | |||
AK214522.1 | - | 1 | 110 | |||
BC010244.1 | - | 10 | 1072 | |||
BC028977.1 | - | 12 | 1430 | 5363085 | ||
BC057296.1 | - | 12 | 1449 | |||
BY551850.1 | - | 1 | 173 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC012634.1 | - | 317 | Chr. - - [-] on Build |