Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ralgps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290570.1 | NP_001277499.1 | 16 | 1724 | ||
NM_001290572.1 | NP_001277501.1 | 17 | 1778 | |||
NM_175211.5 | NP_780420.1 | 19 | 1988 | |||
GenBank mRNA | AK076767.1 | - | 19 | 1973 | ||
AK129123.1 | - | 16 | 1691 | |||
BC037725.1 | - | 5 | 414 | |||
BC072656.1 | - | 17 | 1713 | 30536315 |