Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: Mab21l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172295.4 | NP_758499.3 | 5 | 1043 | ||
GenBank mRNA | AK029040.1 | - | 5 | 782 | ||
AK161057.1 | - | 5 | 960 | |||
AK166928.1 | - | 793 | ||||
AK172389.1 | - | 5 | 992 | |||
BC037703.1 | - | 5 | 980 | 4166596 |