Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033352.3 | NP_001028524.1 | 2 | 1192 | ||
GenBank mRNA | AK052203.1 | - | 2 | 1167 | ||
AK154493.1 | - | 2 | 1193 | |||
BC060201.1 | - | 2 | 1181 | |||
BC141158.1 | - | 2 | 1166 | 9055770 | ||
BC141159.1 | - | 2 | 1166 | 9055771 |