Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gprin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183183.2 | NP_899006.1 | 2 | 689 | ||
GenBank mRNA | AK050148.1 | - | 2 | 690 | ||
AK173336.1 | - | 2 | 693 | |||
BC138149.1 | - | 1 | 559 | 8861168 | ||
BC138150.1 | - | 1 | 559 | 8861169 |