Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033356.3 | NP_001028528.2 | 5, 6 | 1395 | ||
NM_001289692.1 | NP_001276621.1 | 5, 6 | 1395 | |||
NM_001289693.1 | NP_001276622.1 | 6, 7 | 1188 | |||
GenBank mRNA | AK042348.1 | - | 6, 7 | 1190 | ||
AK142898.1 | - | 5, 6 | 1003 | |||
BC150901.1 | - | 5, 6 | 988 | 9087812 | ||
BC172055.1 | - | 5, 6 | 988 |