Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scaper | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081341.1 | NP_001074810.1 | 7 | 621 | ||
GenBank mRNA | AK031358.1 | - | 7 | 607 | ||
AK048755.1 | - | 7 | 628 | |||
AK049219.1 | - | 6 | 585 | |||
AK049497.1 | - | 7 | 617 | |||
AK173175.1 | - | 6 | 569 | |||
BC095992.1 | - | 6 | 569 | |||
BC157959.1 | - | 6 | 584 | 9007286 | ||
BC158062.1 | - | 6 | 561 | 9088165 | ||
BC172119.1 | - | 6 | 584 | 9054405 | ||
BC172151.1 | - | 6 | 561 | 9054821 |