Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101482.2 | NP_001094952.2 | 2 | 149 | ||
NM_001177731.1 | NP_001171202.1 | 3 | 294 | |||
GenBank mRNA | CJ059941.1 | - | 3 | 258 |