Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145156.1 | NP_660138.1 | 2, 3 | 623 | ||
GenBank mRNA | AF377994.1 | - | 2, 3 | 441 | ||
AK155044.1 | - | 2 | 602 | |||
BC025908.1 | - | 2, 3 | 623 | 5043992 |