Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026250.2 | NP_080526.2 | 7 | 1308 | ||
GenBank mRNA | AK014287.1 | - | 7 | 1315 | ||
AK015243.1 | - | 7 | 1287 | |||
AK169361.1 | - | 7 | 1271 | |||
BC033572.1 | - | 6 | 1253 | 5067692 |