Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdsl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133902.2 | NP_598663.2 | 4, 5 | 424 | ||
GenBank mRNA | AF328927.1 | - | 5, 6 | 691 | ||
AK014487.1 | - | 1, 2 | 40 | |||
BC022601.1 | - | 4, 5 | 424 | 4160199 |