Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Axl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190974.1 | NP_001177903.1 | 15 | 2097 | ||
NM_001190975.1 | NP_001177904.1 | 14 | 1833 | |||
NM_009465.4 | NP_033491.2 | 16 | 2124 | |||
GenBank mRNA | AK033237.1 | - | 14 | 2423 | ||
AK155567.1 | - | 17 | 2010 | |||
AK161294.1 | - | 9 | 797 | |||
AK170010.1 | - | 15 | 2071 | |||
AK170897.1 | - | 15 | 1732 | |||
BC008551.1 | - | 11 | 1214 | |||
BC046618.1 | - | 16 | 1961 | 6494383 | ||
BC050914.1 | - | 16 | 1932 | 6418252 | ||
BC058230.1 | - | 16 | 1985 | 6313662 | ||
X63535.1 | - | 16 | 1873 |