Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010661.2 | NP_034791.2 | 4 | 923 | ||
GenBank mRNA | AK053362.1 | - | 4 | 924 | ||
AK143182.1 | - | 4 | 922 | |||
BC131911.1 | - | 4 | 919 | 40130188 | ||
U02880.1 | - | 4 | 920 |
Gene Symbol: A830036E02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK158612.1 | - | 3 | 598 |